missing translation for 'onlineSavingsMsg'
Learn More

Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals

£29.80 - £83.50

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427405
View Documents
Thermo Scientific Alfa Aesar
047132.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15437405
View Documents
Thermo Scientific Alfa Aesar
047132.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-42-5
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7782-42-5
C
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.01
CHEBI:27594
Graphene nanoplatelets aggregates, sub-micronparticles
Black
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Nanoplatelets
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

missing translation for 'einecsNumber' : 231-955-3

missing translation for 'rtecsNumber' : MD9659600

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Graphene nanoplatelet aggregates

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Graphite

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only