missing translation for 'onlineSavingsMsg'
Learn More

Graphite felt, 99.9% (metal basis), Thermo Scientific™

Product Code. 11386938
Click to view available options
Quantity:
10 x 10 cm
25 x 25 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: approx.9.2g/10 x 10cm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00144065

Specifications

Color Gray to Black
Assay Percent Range 99.9% (Metal basis)
Quantity 10 x 10 cm
Chemical Name or Material Graphite felt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'