missing translation for 'onlineSavingsMsg'
Learn More

Graphite flake, median 7-10 micron, 99%, Thermo Scientific Chemicals

£40.90 - £88.04

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11395797
SDS View Documents
Thermo Scientific Alfa Aesar
043480.30
250 g
£40.90
250g
Please to purchase this item. Need a web account? Register with us today!
11305807
SDS View Documents
Thermo Scientific Alfa Aesar
043480.A1
1 kg
£88.04
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H228-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-955-3

missing translation for 'rtecsNumber' : MD9659600

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphite flake, median 7-10 micron, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.