missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
30 x 30 cm
30 x 100 cm
30 x 500 cm
Description
Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray to Black |
Assay Percent Range | 99.8% (Metals basis) |
Quantity | 30 x 500 cm |
Physical Form | Foil |
Chemical Name or Material | Graphite |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'