missing translation for 'onlineSavingsMsg'
Learn More

Graphite foil, 0.5mm (0.02in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

£73.00 - £249.00

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11380228
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042953.VA
15 x 15 cm
£73.00
Each
Please to purchase this item. Need a web account? Register with us today!
11370228
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042953.CG
15 x 60 cm
£102.00
Each
Please to purchase this item. Need a web account? Register with us today!
11390228
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042953.VU
15 x 250 cm
£249.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-42-5
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7782-42-5
C
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.01
CHEBI:27594
Graphite
99.8%, metal basis
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Foil
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-955-3

missing translation for 'rtecsNumber' : MD9659600

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only