Learn More
Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash, Thermo Scientific Chemicals
£30.25 - £72.66
Chemical Identifiers
CAS | 7782-42-5 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
MDL Number | MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301188
|
Thermo Scientific Alfa Aesar
010130.14 |
25 g |
£30.25
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321188
|
Thermo Scientific Alfa Aesar
010130.30 |
250 g |
£40.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311188
|
Thermo Scientific Alfa Aesar
010130.A1 |
1 kg |
£72.66
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-42-5 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7782-42-5 | |
−325 Mesh | |
C | |
UN1325 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
Powder |
Black | |
Odorless | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.01 | |
CHEBI:27594 | |
Graphite powder |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'rtecsNumber' : MD9659600
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Graphite powder, microcrystalline
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only