missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite powder, natural, briquetting grade, -100 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
28 g
113 g
227 g
454 g
Description
Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Black |
Mesh Size | −100 Mesh |
Odor | Odorless |
Quantity | 113 g |
UN Number | UN1325 |
Solubility Information | Insoluble in water. |
Percent Purity | 99.9995% |
Grade | UCP-1 grade, ultra 'F' purity |
Physical Form | Powder |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'