missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite powder, natural, briquetting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
113 g
227 g
454 g
Description
Graphite Powder is basically used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Black |
Mesh Size | −200 Mesh |
Odor | Odorless |
Quantity | 113 g |
UN Number | UN1325 |
Solubility Information | Insoluble in water. |
Percent Purity | 99.9995% |
Grade | UCP-1 grade, ultra 'F' purity |
Physical Form | Powder |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'