Learn More
Graphite powder, natural, universal grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
£43.62 - £476.37
Chemical Identifiers
CAS | 7782-42-5 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
MDL Number | MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375087
|
Thermo Scientific Alfa Aesar
040799.TA |
28 g |
£43.62
28g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11385087
|
Thermo Scientific Alfa Aesar
040799.TB |
113 g |
£130.65
113g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355087
|
Thermo Scientific Alfa Aesar
040799.TC |
227 g |
£231.15
227g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365087
|
Thermo Scientific Alfa Aesar
040799.TD |
454 g |
£476.37
454g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in aircraft manufacturing industries, defense material, automotive industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-42-5 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7782-42-5 | |
−200 Mesh | |
C | |
UN1325 | |
Insoluble. | |
[C] | |
12.01 | |
CHEBI:27594 | |
UCP-1-U grade, ultra 'F' purity | |
(metals basis) |
Black | |
Odorless | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
99.9995% | |
Powder | |
Graphite powder, Natural |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'rtecsNumber' : MD9659600
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only