missing translation for 'onlineSavingsMsg'
Learn More

Graphite powder, nickel coated, -100 mesh, Ni 99.8% (metals basis), Thermo Scientific Chemicals

£34.27 - £104.52

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339288
View Documents
Thermo Scientific Alfa Aesar
043215.22
100 g
£34.27
100g
Please to purchase this item. Need a web account? Register with us today!
11329288
View Documents
Thermo Scientific Alfa Aesar
043215.36
500 g
£104.52
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Conductive surface coatings, inks, adhesives, and sealants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-42-5
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7782-42-5
-100 Mesh
Odorless
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Powder
Black
Ni 99.8% (Metals basis)
C
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.01
CHEBI:27594
Graphite, Nickel Coated
Videos
Safety and Handling

Safety and Handling

Keep away from sources of ignition - No Smoking. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H228-H317-H350-H372

missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only