missing translation for 'onlineSavingsMsg'
Learn More
Graphite powder, synthetic, conducting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals Product Code.: 11395077

Graphite powder, synthetic, conducting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Product Code. 11395077
227 g
Click to view available options
Quantity:
28 g
113 g
227 g
454 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11395077

Brand: Thermo Scientific Alfa Aesar 040797.TC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Graphite powder is basically used for casting polishing purposes and molding. Natural graphite is mostly used for refractories, batteries, steel making, expanded graphite, brake linings, foundry facings and lubricants. It is also used in pigments, anodes arc-lamp carbons, coating cathode ray tubes and moderator in nuclear piles. Graphite (carbon) fiber and carbon nanotubes are also used in carbon fiber reinforced plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size −200 Mesh
Odor Odorless
Quantity 227 g
UN Number UN1325
Solubility Information Insoluble in water as well as other organic solvents.
Percent Purity 99.9995%
Grade UCP-2 grade, ultra 'F' purity
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Graphite powder, Synthetic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN17L018 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog Number040797.TC,040797.TD,040797.TA,040797.TB
Lot NumberX07K002 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number040797.TC,040797.TD,040797.TA,040797.TB
Lot NumberX07K001 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number040797.TC,040797.TD,040797.TA,040797.TB
Lot NumberW04K035 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number040797.TC,040797.TD,040797.TA,040797.TB
Lot NumberW04K036 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number040797.TC,040797.TD,040797.TA,040797.TB

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphite powder, synthetic, conducting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.