missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Graphite rod, 6.15mm (0.242in) dia x 152mm (6in) long, 99.9995% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 014738.KD
280.25 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
This item is not returnable.
View return policy
Description
It is used in various industries. Used in making. Anodes, Electrodes, Heating elements, Crucibles, Hot press dies, Welding plugs, Stirring rods, Lubrication sticks, Bushings & bearings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite rod | |
MFCD00144065 | |
Insoluble in water. |
Tube | |
50 Pc. | |
99.9995% |
RUO – Research Use Only