missing translation for 'onlineSavingsMsg'
Learn More

Graphite tape, square weave, 0.56mm (0.022 in.) thick x 100mm (3.94in) wide, 99.9%, Thermo Scientific™

£78.30 - £257.00

Chemical Identifiers

MDL Number MFCD00144065
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381188
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010136.BU
25 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11371188
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010136.BY
100 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

MFCD00144065

Specifications

MFCD00144065
99.9%
Graphite tape, Square weave, 0.56mm (0.022 in.) thick x 100mm (3.94 in.) wide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-955-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications