missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
Brand: Deutsch Neumann™ 1010129
Code : A0
Additional Details : Weight : 0.02700kg
Description
- Acc. to DIN 12 871
- Durometer 40 Shore A
- Available in a wide range of sizes
Specifications
Gray | |
30mm | |
35 mm | |
Natural Rubber |
Tapered Stopper | |
29 mm | |
1/Ea. |