missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Grey Natural Rubber Stoppers

For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.

Brand:  Deutsch Neumann™ 1010194

 View more versions of this product

Product Code. 10425811

  • £15.15 / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications

Specifications

65mm
Gray
107 mm
Tapered Stopper
1/Ea.
94 mm
Natural Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers