Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
Brand:  Deutsch Neumann™ 1010150
 View more versions of this product
Product Code. 10577941
  • £3.80 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
45mm
Gray
59.5 mm
Tapered Stopper
1/Ea.
50.5 mm
Natural Rubber