Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Accessories
For use with MiniCollect Capillary Blood Collection System
£235.00
Specifications
For Use With (Equipment) Greiner Vacuette MiniCollect Capillary Blood Collection System
View More Specs
Products 1
Product Code Brand Product Type Tube Size dimColor Price Quantity & Availability  
Product Code Brand Product Type Tube Size dimColor Price Quantity & Availability  
13058602
View Documents
Greiner Bio-One
450434
EDTA Tube 80 μL
£235.00
Pack of 1000
Description
Description
  • Capillary or funnel can be inserted into MiniCollect tubes without removing the cap
  • 13 × 75mm carrier tubes facilitate bar code labeling, centrifugation, and storage
  • Use amber carrier tube for sheilding light-sensitive analytes
Specifications
Specifications
Greiner Vacuette MiniCollect Capillary Blood Collection System