missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
For routine clinical chemistry, immunohematology, serology or immunology testing
£229.99 - £306.66
Specifications
Additive | Microscopic Silica Particles with Separation Gel |
---|---|
Material | Plastic |
Closure Type | Non-ridged Pull Cap |
Product Code | Brand | Capacity (Metric) | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Color | Price | Quantity & Availability | ||||
13008652
|
Greiner Bio-One
454067 |
3.5 mL | Red with Yellow Ring |
£229.99
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144534
|
Greiner Bio-One
455071 |
8 mL | Red with Yellow Ring |
£306.66
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15484641
|
Greiner Bio-One
456073 |
13 x 100 mm | Red with Yellow Ring |
£245.31
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13068682
|
Greiner Bio-One
454243 |
2.5 mL | Red with White Ring |
£245.31
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Coated with micronized silica particles which activate clotting when tubes are gently inverted
- Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Pediatric tubes come with white rings for quick identification
- Safety Twist Caps can be removed and replaced several times
- Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
Specifications
Microscopic Silica Particles with Separation Gel | |
Plastic | |
Non-ridged Pull Cap |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator