Learn More
Griffin Education™ Voltmeter with Battery
Brand:  Griffin Education IPC-1505-M
Encompass_Preferred
Product Code. 12733796
  • £49.83 / Each
Explore more special offers
Specifications
1 Pk.
90 mm
Digital
±1 digit, ±0.1% of full scale
130 mm
Voltmeter