Learn More
Kinematica™ Stainless Steel Mixing attachment for MB 800 Laboratory Mixer
Mixing attachment for MB 800 consists of stainless steel, gasket and blade head as well as lid with opening for material addition.
Brand:  Kinematica™ 36050050
 View more versions of this product
Product Code. 11989058
  • £694.00 / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
2000 mL
Stainless Steel
MB 800