Learn More
SPEX™ SamplePrep Grinding Vial Accessory Packages
Contains everything necessary to run Small/Mid-size/Large grinding vials in the LargeFreezer/Mill™
Brand: SPEX™ SamplePrep 6870L
This item is not returnable.
View return policy
Description
- Closed grinding vial prevents cross-contamination.
- Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
- Compatible with grinding vial sets 61043-09 and 61043-11.
- Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Specifications
Grinding Set | |
SamplePrep™ Grinding Vial, Vial Set | |
1 Vial Extractor/Opener (04576-38) and Vial Rack (04576-39) | |
Large |