missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Grip Seal Bags

Provides storage for large number of samples

Brand:  Microspec™ W1

Additional Details : Weight : 1.03000kg

 View more versions of this product

Product Code. 15762175

  • £25.45 / Pack of 1000
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

  • Cost effective sampling
  • Sample storage of large numbers of samples
Specifications

Specifications

Sample Bag
Not Autoclavable
Transparent
Sampling
Non-sterile
Polyethylene
1000
1000
Room Temperature
Grip Seal
Single-use
Polyethylene
50 μm
1000
Box
2 x 9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers