missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Ground Joint Flasks for Vaccum Filtration
Accessories for filter holders for sample preparation.
Brand: Merck XX1514705
252.29 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.Specifications
Clear | |
Standard bottom | |
Borosilicate Glass | |
1 L |