Learn More
Lenz Laborglasinstrumente™ Ground Joint Sleeve with Grip Collar
Product Code. 11542344
Click to view available options
Standard Taper Joint No.:
45/40
Unit Size:
Each
Product Code. 11542344
Brand: Lenz Laborglasinstrumente™ 01101745
Only null left
Add to basket
Add to basket
Sturdy design for permanent use, with grip collar and sealing rings for excellent tightness
  • Outstanding chemical resistance at operating temperatures from -200 to +260 °C
  • With grip collar and sealing rings for excellent tightness
Specifications
For Use With (Equipment) For ground joints NS 45/40
Material PTFE
Quantity 1
Temperature Range (Metric) -200 to +260 °C
Standard Taper Joint No. 45/40