missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Mouse, Clone: GH/1450, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2688-MSM3-P0
This item is not returnable.
View return policy
Specifications
Growth Hormone | |
Cytoplasmic | |
GH/1450 | |
Unconjugated | |
NP_072053 | |
Mouse | |
20kDa | |
Protein A/G Purified | |
RUO | |
2688 | |
IgG2b |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
GH1 | |
A recombinant human Growth Hormone (GH) fragment (aa58-187) | |
Pituitary cells or Pituitary | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |