missing translation for 'onlineSavingsMsg'
Learn More

Growth Hormone Mouse, Clone: GH/1450, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2688-MSM3-P1

 View more versions of this product

Product Code. 15970088

  • £395.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Growth Hormone
Cytoplasmic
GH/1450
Unconjugated
NP_072053
Mouse
20kDa
Protein A/G Purified
RUO
2688
IgG2b
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
GH1
A recombinant human Growth Hormone (GH) fragment (aa58-187)
Pituitary cells or Pituitary
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers