missing translation for 'onlineSavingsMsg'
Learn More

GSK 269962, Tocris Bioscience™

Product Code. 18076022
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective ROCK inhibitor

Potent Rho kinase (ROCK) inhibitor (IC50 values are 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively). Displays greater than 30-fold selectivity for ROCK against a panel of serine/threonine kinases. Induces vasorelaxation in preconstricted rat aorta (IC50 = 35 nM); lowers blood pressure in a rat model of hypertension.

Chemical Identifiers

CAS 850664-21-0
Molecular Formula C29H30N8O5
Molecular Weight (g/mol) 570.61
InChI Key YOVNFNXUCOWYSG-UHFFFAOYSA-N
Synonym aminofurazanyl-azabenzimidazole 6n, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-morpholin-4-yl ethoxy benzamide, kinome_3843, d02zqs, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethoxy benzamide, n-3-2-4-amino-furazan-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethyl oxy benzamide, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethylimidazo 5,4-d pyridin-6-yl oxyphenyl-4-2-morpholin-4-ylethoxy benzamide
PubChem CID 16095342
IUPAC Name N-[3-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-6-yl]oxyphenyl]-4-(2-morpholin-4-ylethoxy)benzamide
SMILES CCN1C2=CC(=NC=C2N=C1C3=NON=C3N)OC4=CC=CC(=C4)NC(=O)C5=CC=C(C=C5)OCCN6CCOCC6

Specifications

Quantity 50 mg
Formula Weight 570.6
Percent Purity >99%
Chemical Name or Material GSK 269962
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'