missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 6853
Potent and selective BRPF1 inhibitor
£71.27 - £268.89
Specifications
Chemical Name or Material | N-[2,3-dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-2-methoxybenzamide |
---|---|
CAS | 1910124-24-1 |
Molecular Formula | C22H27N5O3 |
Formula Weight | 409.48 |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785716
|
Tocris Bioscience™
6198/25 |
25 mg |
£268.89
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18726046
|
Tocris Bioscience™
6198/5 |
5 mg |
£71.27
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Potent and selective BRPF1 inhibitor
- Exhibits >1600-fold selectivity for BRPF1 over other bromodomains (pIC50 values by TR-FRET are 8.1, 5.1/4.8 and 4.7/<4.3 for BRPF1, BRPF2/3 and BRD4 BD1/2, respectively)
- Negative Control also available
Specifications
N-[2,3-dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-2-methoxybenzamide | |
C22H27N5O3 | |
98% |
1910124-24-1 | |
409.48 |
Safety and Handling
Recommended Storage : Store at -20°C