missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GSK-J4 hydrochloride, 95%
Brand: Thermo Scientific™ 468001000
573.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
GSK-J4 hydrochloride | |
Ethyl 3-((6-(4,5-dihydro-1 H-benzo[d]azepin-3(2 H)-yl)-2-(pyridin-2-yl)pyrimidin-4-yl)amino)propanoate hydrochloride | |
C24 H27 N5 O2. H Cl | |
Solid | |
95% |
1797983-09-5 | |
100 mg | |
453.96 | |
Reagent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
344444 | Certificate of Analysis | 19/10/2020 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ GSK-J4 hydrochloride, 95% > 100 mg
Spot an opportunity for improvement?Share a Content Correction