Learn More
Fisherbrand™ GT1 Benchtop Centrifuge Packs
Fisherbrand™ Benchtop Centrifuges combine a compact size with operational flexibility, making them the ideal choice for molecular biology laboratories. Available in ventilated or refrigerated versions.
£3576.84 - £6000.96
Specifications
Certifications/Compliance IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, UL Listed, CE marked, Certified biocontainment
Capacity 4 x 145 mL
Max. RCF 3,046 x g
Max. Speed 4,350 rpm
Drive System Direct Brushless Induction
View More Specs
Products 2
Product Code Brand Model Refrigerated Price Quantity & Availability  
Product Code Brand Model Refrigerated Price Quantity & Availability  
15868722
View Documents Promotion Details
Fisherbrand™
50156859
GT1R Yes
£6000.96
Each
15878722
View Documents Promotion Details
Fisherbrand™
50156848
GT1 No
£3576.84
Each
Description
Description
  • Compact size with minimal benchtop footprint
  • Ventilated and refrigerated versions available, with rapid temperature pull-downs
  • Includes TX-150 swing-out rotor with adapters accommodating 4x50mL or 8x15mL conical tubes
  • One-touch, easy-to-use operation with pre-saved protocols
  • Backlit display interface with multi-language options
  • Rapid push-button rotor exchange with the Auto-Lock™ rotor swap system
  • Corrosion resistant and lubricating rotor-cross coating
  • Fitted with ClickSeal™ biocontainment lids
  • Optional high-speed microlitre rotor also available for molecular biology applications
  • Glove and wipe-clean, detergent-friendly
  • Two year warranty
Specifications
Specifications
IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, UL Listed, CE marked, Certified biocontainment
3,046 x g
Direct Brushless Induction
Four direct access programs
Yes
ClickSeal biocontainment lid
2 Years (Centrifuge), 5 Years (Rotor)
50/60 Hz
4 x 145 mL
4,350 rpm
TX-150 rotor, ClickSeal lid, four adapters for 4x50mL conical tubes, four adapters for 8x15mL conical tubes
TX-150
10 sec. to 99 hr. 59 min.
Benchtop Centrifuge
230 V
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ GT1 Benchtop Centrifuge Packs