Learn More
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry)
£23.20 - £785.00
Chemical Identifiers
CAS | 50-01-1 |
---|---|
Molecular Formula | CH5N3·HCl |
Molecular Weight (g/mol) | 95.53 |
InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
Synonym | guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride |
PubChem CID | 5742 |
ChEBI | CHEBI:32735 |
IUPAC Name | guanidine;hydrochloride |
SMILES | C(=N)(N)N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10132340
|
Thermo Scientific Acros
120232500 |
250 g | Plastic bottle |
£23.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10447690
|
Thermo Scientific Acros
120230010 |
1 kg | Plastic bottle |
£60.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337180
|
Thermo Scientific Acros
120230025 |
2.5 kg | Plastic Bottle |
£110.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10573174
|
Thermo Scientific Acros
120230100 |
10 kg | Plastic drum |
£310.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965591
|
Thermo Scientific Acros
120230251 |
25 kg | Fibre drum |
£785.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50-01-1 | |
95.53 | |
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
CHEBI:32735 | |
C(=N)(N)N.Cl |
CH5N3·HCl | |
PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
5742 | |
guanidine;hydrochloride |
Specifications
50-01-1 | |
White to Yellow | |
Authentic | |
CH5N3·HCl | |
03,82 | |
1.34 | |
Solubility in water: 2280g/L (20°C). Other solubilities: 300g/L ethanol (30°C), 19.5g/100g dimethylformamide (30°C), <0.05g/100g acetone -<0.05g/100g benzene (30°C), insoluble in trichloroethylene | |
C(=N)(N)N.Cl | |
95.53 | |
CHEBI:32735 | |
98% | |
Guanidine hydrochloride |
179.0°C to 186.0°C | |
1.3400g/mL | |
0.5% max. Ammonium chloride (Argentometry) | |
H2NC(=NH)NH2·HCl | |
15,4597 | |
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
guanidine;hydrochloride | |
5742 | |
95.53 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-002-3
- Guanidine hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only