missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, ≥99.5%, (Crystalline/Ultra Pure), MP Biomedicals

Product Code. 11442002
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Forms clear, colorless solutions at 6.0 M concentration. O.D. 260 nm of a 6.0 M solution: <0.03

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Assay Percent Range ≥99.5%
Quantity 100 g
Formula Weight 95.5
Grade Ultrapure
Physical Form Crystalline
Chemical Name or Material Guanidine Hydrochloride
compliance-icons
Product Identifier
  • Guanidine Hydrochloride, Ultra Pure
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'