missing translation for 'onlineSavingsMsg'
Learn More
Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™ Product Code.: 10503345

Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™

Product Code. 10503345
1 kg, Poly Bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy

Product Code. 10503345

Brand: Fisher BioReagents BP2211

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the isolation of intact RNA from cell extracts

A more powerful denaturing agent than Guanidine Hydrochloride, Guanidine Thiocyanate is suitable for the isolation of intact RNA from cell extracts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-84-0
Molecular Formula C2H6N4S
Synonym Guanidinium Isothiocyanate

Specifications

Absorbance 0.8 max. (70% in water at 25°C) at 280nm
Melting Point 118°C
Color White
pH 5
Assay Percent Range ≥99.0 %
Packaging Poly Bottle
Linear Formula H2NC(:NH)NH2·HSCN
Quantity 1 kg
Solubility Information Soluble in water
Formula Weight 118.16
Vapor Pressure Negligible
Percent Purity ≥99.0%
Physical Form Powder or Solid
Chemical Name or Material Guanidine Thiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Guanidine thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.