missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LRRTM2 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-41129PEP
Additional Details : Weight : 0.00970kg
Description
A blocking peptide containing 17 amino acid peptide near the carboxy terminus of human LRRTM2. Source: Synthetic (Accession #: EAW62125) The LRRTM2 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
26045 | |
Immunogen affinity purified | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP2-41129. For Further Blocking Peptide Related Protocol, Click Here . | |
LRRTM2 |
Human | |
LRRTM2 | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin | |
leucine-rich repeat transmembrane neuronal protein 2, leucine rich repeat transmembrane neuronal 2 | |
50 μg |