missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Guanosine-5'-diphosphate disodium salt Product Code.: 15404209

Thermo Scientific Chemicals Guanosine-5'-diphosphate disodium salt

Product Code. 15404209
100 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 15404209

Brand: Thermo Scientific Chemicals J61646.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guanosine-5'-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine-5′-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.

Solubility
Soluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7415-69-2
Molecular Formula C10H13N5Na2O11P2
Molecular Weight (g/mol) 487.17
MDL Number MFCD00066538
InChI Key LTZCGDIGAHOTKN-KHRSEZDTNA-L
Synonym 5-gdp pound not2na
PubChem CID 134129638
IUPAC Name disodium;(2R,3S,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water
Formula Weight 487.17
Physical Form Powder
Chemical Name or Material Guanosine-5'-diphosphate disodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14L517 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog NumberJ61646.MC,J61646.ME
Lot NumberP14L516 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog NumberJ61646.MC,J61646.ME
Lot NumberX29K530 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberJ61646.MC,J61646.ME
Lot NumberU25K536 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberJ61646.MC,J61646.ME
Lot NumberQ19K515 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ61646.MC,J61646.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Guanosine-5'-diphosphate disodium salt >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.