missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Description
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.
Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.
Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.
Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.
Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | ∼250°C (decomposition) |
Quantity | 25 g |
Beilstein | 625911 |
Merck Index | 14,4566 |
Solubility Information | Soluble in Formic Acid:Water (1:1): 50mg/mL. Insoluble in methanol,diethyl ether. |
Optical Rotation | −72° (c=1.5 in 0.1N NaOH) |
IUPAC Name | 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
Formula Weight | 283.24 |
Percent Purity | ≥98% |
Chemical Name or Material | Guanosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'