Learn More
OHAUS™ Guardian 7000 Hotplate-Stirrer
Durable hotplate stirrers with superior safety features and intelligent performance
Brand: OHAUS™ 30500613
1484.95 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Protect your lab with patented SmartPresence™ & SmartLink™ technologies. These exclusive features automatically shut off the heater if no one is detected after a user set "time out" period has passed.
- An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
- Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
Specifications
Ceramic | |
5.443 kg | |
e-G71HS10C | |
123.000 mm | |
4.80 in. | |
286.00 mm | |
11.25 in. | |
5.200 kg |
230 V | |
230 V, 7 A, 50/60 Hz | |
16.30 in. | |
+ 5°C – 500°C | |
50/60 Hz | |
415.00 mm | |
11.500 lb | |
EU |