missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Guinea Pig IL-10 DuoSet ELISA
Sufficient for at least 15 96-well plates
Marke: R&D Systems™ DY5258
Weitere Details : Weight : 0.09700kg
Beschreibung
The Guinea Pig IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies guinea pig IL-10.Spezifikation
Biotin | |
ELISA | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
62.50 to 4000 pg/mL | |
100135553 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |