missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Guvacine hydrochloride, 99+%
Click to view available options
Quantity:
100 mg
Specifications
Specifications
CAS | 498-96-4 |
Color | White |
Molecular Formula | C6H9NO2 |
MDL Number | MFCD00055191 |
Quantity | 100 mg |
Merck Index | 14,4584 |
Synonym | guvacine, 1,2,5,6-tetrahydropyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-3-pyridinecarboxylic acid, 1,2,5,6-tetrahydronicotinic acid, 1,2,5,6-tetrahydro-pyridine-3-carboxylic acid, spectrum_001426, tocris-0234, specplus_000808, lopac-g-007, spectrum2_001474 |
InChI Key | QTDZOWFRBNTPQR-UHFFFAOYSA-N |
SMILES | C1CNCC(=C1)C(=O)O |
IUPAC Name | 1,2,3,6-tetrahydropyridine-5-carboxylic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'