missing translation for 'onlineSavingsMsg'
Learn More

GW3965 hydrochloride, MedChemExpress

MedChemExpress GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.

£82.28 - £919.60

Specifications

Chemical Name or Material GW3965 hydrochloride
CAS 405911-17-3
Color White
Purity Grade Notes Research
Molecular Formula C33H32Cl2F3NO3
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16484078
Encompass_Preferred
View Documents
Medchem Express
HY-10627A
10 mg
£136.73
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16430437
Encompass_Preferred
View Documents
Medchem Express
HY-10627A
50 mg
£536.03
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16415056
Encompass_Preferred
View Documents
Medchem Express
HY-10627A
100 mg
£919.60
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16406787
Encompass_Preferred
View Documents
Medchem Express
HY-10627A
5 mg
£82.28
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.
Specifications

Specifications

GW3965 hydrochloride
White
C33H32Cl2F3NO3
ClC1=C(C(F)(F)F)C=CC=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.[H]Cl
618.51
Research
COVID-19-immunoregulation
405911-17-3
Research
DMSO : 100 mg/mL (161.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
618.51
98.13%
Solid
Videos
Safety and Handling

Safety and Handling

H302∣H318∣H413

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.