Learn More
Thermo Scientific™ H-12000 Bioprocessing Swinging-Bucket Rotor
Integrate the Thermo Scientific™ H-12000 Bioprocessing Swinging-Bucket Rotor with Thermo Scientific™ Sorvall™ RC12BP low-speed centrifuges.
Brand:  Thermo Scientific™ 77080
Product Code. 10308369
  • £19500.00 / Each
Explore more special offers
Description
Description
Integrate the Thermo Scientific™ H-12000 Bioprocessing Swinging-Bucket Rotor with Thermo Scientific™ Sorvall™ RC12BP low-speed centrifuges.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
H-12000 Bioprocessing Swinging-Bucket Rotor
Swinging Bucket Rotor
7,340 x g
297.2 mm
8117
90°
Stainless Steel Rotor Body, Aluminum Bucket
6 x 2400 mL
4,700 rpm
141 mm
Thermo Scientific Sorvall RC 12 Series Large Capacity Centrifuges
3,610 x g