Description
Achieve various capacities with adapters ranging from 300mL to 2,000mL by using the Thermo Scientific™ H-12000 Blood Processing Swinging-Bucket Rotor.
Compatible with:Thermo Scientific™ Sorvall™ RC12BP low-speed centrifuges
Specifications
H-12000 Blood Processing Swinging-Bucket Rotor | |
Swinging Bucket Rotor | |
7,340 x g | |
297.2 mm | |
8117 | |
90° |
Stainless Steel Rotor Body, Aluminum Bucket | |
12 x 500 mL | |
4,700 rpm | |
146.3 mm | |
Thermo Scientific Sorvall RC 12 Series Large Capacity Centrifuges | |
3,610 x g |