Learn More
SP Bel-Art™ H-B DURAC Liquid Crystal Body Forehead Thermometer 100/Pk.
Non-invasive body temperature measurement is ideal for workforce personnel.
Brand:  SP Bel-Art™ B62010-0100
Product Code. 16626652
  • £227.00 / Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Color-coded readings allow for quick evaluation of human temperature
  • Easy to use - simply peel off backing and stick to forehead or hand
  • Can be self-applied to maintain social distancing during temperature testing
  • Latex-free
  • Utilizes Thermocratic Liquid Crystal technology
  • Disposable
  • Instructions included
Specifications
Specifications
Single-use
45 mm
40°C
34°C