missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion
Individual Calibration Report as needed for many standard operating procedures
£247.00 - £290.00
Specifications
Certifications/Compliance | Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA |
---|---|
For Use With (Application) | Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. |
Grade | Precision |
Immersion Type | Partial Immersion |
Material | Glass |
Product Code | Brand | Temperature Range (Metric) | Accuracy | Subdivision | Calibration Points | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Accuracy | Subdivision | Calibration Points | Price | Quantity & Availability | ||
17521535
|
H-B Instrument™
B60202-0100 |
-20°C to +110°C | ±1°C | 1°C | 0°, 50°, 100°C |
£247.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||
13069142
|
H-B Instrument™
B60202-0400 |
-20°C to +150°C | ±1°C | 1°C | 0°, 60°, 120°C |
£290.00
Each |
In Stock
Log in to see stock. |
||
13049142
|
H-B Instrument™
B60202-0000 |
-10°C to +70°C | ±0.5°C | 0.5°C | 0°, 37°, 56°C |
£267.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||
Description
- Fill: Black spirit
- Each thermometer is individually calibrated
- Celsius-scale
- Designed to be very easy to read
- Replace mercury thermometers with the same accuracy without any hazards
- Nontoxic, nonhazardous, biodegradable
- NIST-Certified detailed, laminated report of calibration
- Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read
- Individually calibrated to NIST standards at specific temperature and supplied with NIST report of calibration stating corrections to one-tenth the smallest division
- Contain an environmentally friendly non-mercury liquid that is non-toxic, non-hazardous, and fully biodegradable
- In case of breakage the liquid can be cleaned up easily with soap and water
- “Lead-free”, yellow back glass and black liquid provides easy readability and fewer reading errors
- Contains non-toxic, biodegradable, Enviro-Safe™ liquid
- EnviroKleen™ certified
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Certifications
Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST™ standards used for the calibration. Individually serialized and made in the USA.
Specifications
Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA | |
Precision | |
Glass | |
76 mm |
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. | |
Partial Immersion | |
Enviro-Safe™ Liquid |