missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers
GREENER_CHOICE

Product Code. p-4555064 Shop All SP Scienceware Products
Click to view available options
Material:
PFA-coated
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13596980

Brand: H-B Instrument™ B605100100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

EnviroKleen™-certified environmentally friendly with 100% non-toxic, non-hazardous, biodegradable liquid fill. H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers are 175mm (6.9 in.) liquid-in-glass, 35mm immersion thermometers.

  • USA made thermometers include Enviro-Safe™ green liquid against white back glass
  • EnviroKleen™-certified environmentally friendly and packaged in recyclable Safety-Pak™ packaging
  • Feature the same accuracy as similar mercury thermometers, with arrow indicators imprinted on the glass at 25, 30, 37 and 56°C
  • Individually serialized and tested in a triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory
  • Internationally recognized and accepted by the signatory economies of the ILAC/MRA
  • Multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
  • Mfr. No. B60510-0100 features PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage 

Specifications

Certifications/Compliance ISO/IEC 17025:2005, A2LA, NIST
Length (English) Overall 6.9 in.
Length (Metric) Overall 175 mm
Material PFA-coated
Thermometer Fill Enviro-Safe™ Liquid
Product Type Dry Block/Incubator Thermometer
Max. Temperature (Metric) 57°C
Min. Temperature (Metric) 24°C
Temperature Range (Metric) 24°C to 57°C
Accuracy 0.5°C
Subdivision 0.5°C
Immersion Length 35 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.