missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ FRIO-Temp™ Calibrated Thermometer with Toluene Fill

Calibrated thermometer with a PFA-fluoropolymer safety coating

Brand:  H-B Instrument™ B60200-0500

Additional Details : Weight : 0.11570kg Transport : UN number : 1090 Chem class : 3 Pack group : II

Product Code. 17531275

  • £311.00 / Each
Estimated Shipment: 02-10-2024
to see stock.

Description

Description

With toluene fill and glass bead-filled bottle, designed for use in ultralow freezers.

Supplied with a detailed calibration report, this thermometer is certified and traceable to NIST and DAkkS.

  • Type: Calibrated; certified to 0.1 of the smallest division
  • Coating: Fluoropolymer coated for safety
  • Thermometer fill:Toluene
  • Bottle Fill: Glass Beads
  • Divisions: 1
  • Accuracy: 1°C, 2° from -1 to -40°C; 2.5°C below -40°C
  • Certification Point: -40°C
  • Individually serialized

Specifications

Specifications

ISO/IEC 17025:2005, A2LA, NIST, DAkkS and FRIO-Temp™ Certified
5.7 in.
Glass
125°C
-90°C to +125°C
Freezer, refrigerator, incubator and ovens to verify sample temperature
145 mm
Toluene
-90°C
2°C from -1 to -40°C, 2.5°C below -40
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers