missing translation for 'onlineSavingsMsg'
Learn More

Labcon H-Pette™ For Blood Smear Preparation

This dispenser makes blood slide preparation safe and easy. By not having to open the primary sample tube, you greatly reduce the risk of lab-acquired infection by minimizing aerosols and exposure to blood borne pathogens.

Brand:  Labcon 7620-500-010-9

73.18 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 17404282

  • £108.00 / Pack of 500

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

H-Pette™ simplifies blood smear preparation by cutting out several steps to improve efficiency, saving time and reducing wasted materials or blood samples. Using the H-Pette™ eliminates the need for sticks, capillaries, or syringes to dispense blood on the slide. No need for special training, fittings, or inserts.

  • Easy delivery of blood sample to a glass slide
  • Minimizes aerosols and reduces the risk of lab-acquired infections
  • No special training, special fittings or inserts
  • Fits all standard 13 mm and 16 mm glass or plastic blood tubes with rubber stoppers
  • Polystyrene
  • Not Autoclavable
  • Made in USA
  • Cannula tip length is 0.18 in.
Specifications

Specifications

Blood Droplet Dispenser
Single-use
500 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers