missing translation for 'onlineSavingsMsg'
Learn More

Labcon H-Pette™ For Blood Smear Preparation

This dispenser makes blood slide preparation safe and easy. By not having to open the primary sample tube, you greatly reduce the risk of lab-acquired infection by minimizing aerosols and exposure to blood borne pathogens.

Brand:  Labcon 7620-540-000-9

 View more versions of this product

Product Code. 17474362

  • £28.60 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

H-Pette™ simplifies blood smear preparation by cutting out several steps to improve efficiency, saving time and reducing wasted materials or blood samples. Using the H-Pette™ eliminates the need for sticks, capillaries, or syringes to dispense blood on the slide. No need for special training, fittings, or inserts.

  • Easy delivery of blood sample to a glass slide
  • Minimizes aerosols and reduces the risk of lab-acquired infections
  • No special training, special fittings or inserts
  • Fits all standard 13 mm and 16 mm glass or plastic blood tubes with rubber stoppers
  • Polystyrene
  • Not Autoclavable
  • Made in USA
  • Cannula tip length is 0.18 in.
Specifications

Specifications

Blood Droplet Dispenser
Single-use
100 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Product Guides

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labcon H-Pette™ For Blood Smear Preparation > H-Pette Dispenser; Product Type: Blood Droplet Dispenser; Disposable: Yes; Quantity: 100 Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.