Learn More
3M™ H422000 Battery
3M™ H422000 Battery is a lithium 5V battery for use with Speedglas™ Welding Filters.
Brand: 3M™ 7100167881
6.90 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.47900kg Transport : UN number : 3090 Chem class : 9
Description
- Lithium 5V batteries for welding filters
Specifications
Lithium 5 V | |
Silver | |
Speedglas Welding Filters 9100/SL/100 | |
Battery |