missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Monoclonal Antibody (12CA5), Agarose, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112455
Code : LD
Additional Details : Weight : 0.01000kg
Description
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.Specifications
HA Tag | |
Monoclonal | |
Agarose | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
A 9-amino acid peptide corresponding to influenza virus hemagglutinin | |
1 mL | |
Primary | |
4° C, do not freeze | |
Slurry | |
Tag |
Immunoprecipitation | |
12CA5 | |
PBS with 0.08% sodium azide | |
Mouse | |
Protein A/G | |
RUO | |
Tag | |
Antibody | |
IgG2b |